Learn More
Sigma Aldrich Fine Chemicals Biosciences fetal Bovine Serum USDA ap
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F092650ML
Encompass
fetal bovine serum (fBS) or fetal calf serum is the blood fraction remaining after the natural coagulation of blood. It comes from the blood drawn from a bovine fetus via a closed system of collection at the slaughterhouse. It is the most widely used serum-supplement for the in vitro cell culture of eukaryotic cells.
Catalog No. 50-290-1833