Learn More
Bioworld Fetal bovine serum - premium grade, 500 mL
Catalog No. 50254076
Encompass_Preferred
Please inquire for availability and specifications.Triple 0.1µm sterile-filteredMycoplasma-tested and virus-screenedShips on dry ice. Store frozen at -20 to -10°C
Catalog No. 50-254-076 Supplier Bioworld Supplier No. 306110281
Please inquire for availability and specifications.Triple 0.1µm sterile-filteredMycoplasma-tested and virus-screenedShips on dry ice. Store frozen at -20 to -10°C