Learn More
Simmler Fetal Cell Stain Kit 100 Tests
Catalog No. NC9775723
Encompass_Preferred
Fetal Cell Stain Kit; Convenient flip-open spout caps; Provide complete staining in less than 20 min.; Includes one 120ml bottle of each reagent, Fixing Solution, Buffer Solution and the Fetal Cell Staining Solution; Exp: 18month; 100/PK
Catalog No. NC9775723 Supplier Simmler Supplier No. S0412100
Fetal Cell Stain Kit; Convenient flip-open spout caps; Provide complete staining in less than 20 min.; Includes one 120ml bottle of each reagent, Fixing Solution, Buffer Solution and the Fetal Cell Staining Solution; Exp: 18month; 100/PK