Learn More
SURE TECH DIAGNOSTIC ASSOC INC Fetal Hemoglobin Controls (2 sets)
Catalog No. NC0502894
Encompass_Preferred
Fetal Hemoglobin Controls . Each set includes three levels of controls and one vials of each level. These whole blood controls are stored refrigerated and have a 90 day out date. They are manufactured quarterly in January, April, July and October.
Catalog No. NC0502894 Supplier SURE TECH DIAGNOSTIC ASSOC INC Supplier No. ST 201
Fetal Hemoglobin Controls . Each set includes three levels of controls and one vials of each level. These whole blood controls are stored refrigerated and have a 90 day out date. They are manufactured quarterly in January, April, July and October.