Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Fetal Pig Dissection 3D Model Making Kit
SCI_ED

Catalog No. S436047 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S436047 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S436047 Supplier NewPath Learning™ Supplier No. 247740
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Fetal Pig Dissection 3D Model Making Kit allows students to build a model fetal pig with organs to learn about the anatomy of the major body systems.

  • Through the process of building a model fetal pig, they build the pig and then add the organs to the model.
  • The model can be used look to at the anatomy of the major body systems and covers the nervous system, circulatory system, respiratory system, digestive system, excretory system, and male and female reproductive systems.
  • The model also shows how a fetal pig differs from an adult animal through coverage of fetal circulation and the umbilical cord.
  • Students can then carry out a dissection to see how the parts fit together for comparative anatomy as well as an excellent means to prepare for a dissection.
  • The Model Making Kit includes enough materials for students to assemble up to 5 paper models working cooperatively.
  • The comprehensive Teacher Resource Guide included provides illustrated readings, activities, key vocabulary and an assessment, along with teaching opportunities for students to Engage, Explore, Explain, Extend and Evaluate their understanding of the concepts presented.

Specifications

For Use With (Application) Model Assembly
Grade 6 to 10
Includes 5 Models, Teacher Resource Guide, Vinyl Pouch
Length (English) 12 in.
Material Paper
Weight (English) 2 lb.
Width (English) 9 in.
Product Type 3D Model
Grade Level Intermediate - Middle School, High School
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.