Learn More
SURE TECH DIAGNOSTIC ASSOC INC Fetal Hemoglobin Controls (1 sets)
Catalog No. NC0866249
Encompass_Preferred
Fetal Hemoglobin Controls . Each set includes three levels of controls and two vials of each level. These whole blood controls are stored refrigerated and have a 90 day out date. They are manufactured quarterly in January, April, July and October.
This item is not returnable. View return policy
Catalog No. NC0866249 Supplier SURE TECH DIAGNOSTIC ASSOC INC Supplier No. ST 201B
 Restricted Item
Add to Cart
Add to Cart
Fetal Hemoglobin Controls . Each set includes three levels of controls and two vials of each level. These whole blood controls are stored refrigerated and have a 90 day out date. They are manufactured quarterly in January, April, July and October.