Learn More
Aviva Systems Biology Corp FETUB Antibody - N-terminal region (ARP42400_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42400_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FETUB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FETUB. Alternate names: 16G2, Gugu, IRL685ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58213