Learn More
New England Biolabs, Inc. Fetuin – 500 µg
SDP
Supplier:  New England Biolabs, Inc. P6042S
Encompass
Fetuin is a glycoprotein containing sialylated N-linked and O-linked glycans that can be used as a positive control for endoglycosidase enzymes that cleave both N-linked and O-linked carbohydrates.
Catalog No. 50-591-179