Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fetuin A (human), recombinant, expressed in HEK293 cells | 9014-81-7 | >95.0% | 500UG
SDP

Catalog No. 5000176641
Encompass_Preferred

Recombinant human Fetuin A (AHSG) produced in HEK293 cells with a C-terminal His tag. Supplied as a lyophilized powder formulated in 20 mM PB, 150 mM NaCl, pH 7.4, the product is intended for research applications requiring a high-purity serum glycoprotein involved in mineralization and cellular processes.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • High purity (>95% by reducing SDS-PAGE).
  • Approximate molecular weight ~60.0 kDa (349 aa, A19-V367).
  • Lyophilized formulation in 20 mM PB, 150 mM NaCl, pH 7.4.
  • Recommended storage at -20°C; aliquot and freeze at -80°C for long-term storage.
  • Reported biological activity: inhibits calcium phosphate precipitation (IC50 46.163 μg/mL).

Catalog No. 50-001-76641 Supplier Medchemexpress LLC Supplier No. HYP70170500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Fetuin A (AHSG) produced in HEK293 cells with a C-terminal His tag. Supplied as a lyophilized powder formulated in 20 mM PB, 150 mM NaCl, pH 7.4, the product is intended for research applications requiring a high-purity serum glycoprotein involved in mineralization and cellular processes.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • High purity (>95% by reducing SDS-PAGE).
  • Approximate molecular weight ~60.0 kDa (349 aa, A19-V367).
  • Lyophilized formulation in 20 mM PB, 150 mM NaCl, pH 7.4.
  • Recommended storage at -20°C; aliquot and freeze at -80°C for long-term storage.
  • Reported biological activity: inhibits calcium phosphate precipitation (IC50 46.163 μg/mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.