Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fetuin B protein, mouse | >95.0% | 10 UG
SDP

Catalog No. 5000293613
Encompass_Preferred

Recombinant mouse Fetuin B protein produced in HEK293 cells and supplied lyophilized with a His tag. The material is intended for research use in biochemical assays, binding studies, and functional characterization, offering high purity and low endotoxin to support sensitive assays. Storage and reconstitution recommendations are provided by the manufacturer.

  • Recombinant mouse Fetuin B expressed in HEK293 cells.
  • His tag for affinity purification.
  • Apparent molecular weight 55-65 kDa.
  • Purity >95.0% by SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Lyophilized from 20 mM Tris, 150 mM NaCl, pH 7.5.
  • Store lyophilized at -20°C; aliquot and freeze after reconstitution.

Catalog No. 50-002-93613 Supplier Medchemexpress LLC Supplier No. HYP7519510UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse Fetuin B protein produced in HEK293 cells and supplied lyophilized with a His tag. The material is intended for research use in biochemical assays, binding studies, and functional characterization, offering high purity and low endotoxin to support sensitive assays. Storage and reconstitution recommendations are provided by the manufacturer.

  • Recombinant mouse Fetuin B expressed in HEK293 cells.
  • His tag for affinity purification.
  • Apparent molecular weight 55-65 kDa.
  • Purity >95.0% by SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Lyophilized from 20 mM Tris, 150 mM NaCl, pH 7.5.
  • Store lyophilized at -20°C; aliquot and freeze after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.