Learn More
Medchemexpress LLC HY-P2352 50mg Medchemexpress, Fetuin, Fetal Bovine Serum CAS:9014-81-7 Purity:>98%
SDP
Catalog No. 502083087
Encompass_Preferred
Medchemexpress, HY-P2352 50mg Fetuin, Fetal Bovine Serum CAS:9014-81-7 Fetuin, Fetal Bovine Serum is a liver-secreted 64 kDa plasma glycoprotein isolated from fetal bovine serum. Fetuin, Fetal Bovine Serum inhibits trypsin activity and promote cellular attachment, growth, and differentiation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3087 Supplier Medchemexpress LLC Supplier No. HYP235250MG
Medchemexpress, HY-P2352 50mg Fetuin, Fetal Bovine Serum CAS:9014-81-7 Fetuin, Fetal Bovine Serum is a liver-secreted 64 kDa plasma glycoprotein isolated from fetal bovine serum. Fetuin, Fetal Bovine Serum inhibits trypsin activity and promote cellular attachment, growth, and differentiation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.