Learn More
Selleck Chemical LLC Fetuin Fetal Bovine Serum P1243-100MG
SDP
Catalog No. 5000039397
Encompass_Preferred
Fetuin Fetal Bovine Serum a prevalent protein found in fetal bovine plasma is the bovine equivalent of human 2HS glycoprotein ( 2HS) commonly used as a supplement in various serum-free media It inhibits trypsin activity and enhances cellular attachment growth and differentiation in culture systems
Catalog No. 50-000-39397 Supplier Selleck Chemical LLC Supplier No. P1243100MG
Fetuin Fetal Bovine Serum a prevalent protein found in fetal bovine plasma is the bovine equivalent of human 2HS glycoprotein ( 2HS) commonly used as a supplement in various serum-free media It inhibits trypsin activity and enhances cellular attachment growth and differentiation in culture systems