Learn More
Medchemexpress LLC Fetuin, Fetal Bovine Serum | 9014-81-7 | 95% | 64 kDa | 250 MG
SDP
Catalog No. 5000426219
Encompass_Preferred
Fetuin, Fetal Bovine Serum is a liver-secreted 64 kDa plasma glycoprotein isolated from fetal bovine serum. It inhibits trypsin activity and promotes cellular attachment, growth, and differentiation. It is often an important supplement to many serum-free media. This acidic glycoprotein features three N-linked and three O-linked oligosaccharide chains, with terminal sugar residues rich in sialic acid (N-acetylneuraminic acid), contributing to its net negative charge.
  • Inhibits trypsin activity
  • Promotes cellular attachment
  • Promotes growth
  • Promotes differentiation
  • Important supplement to many serum-free media
  • Acidic glycoprotein with N-linked and O-linked oligosaccharide chains
  • Terminal sugar residues rich in sialic acid
Catalog No. 50-004-26219 Supplier Medchemexpress LLC Supplier No. 250MGHYP2352
Add to Cart
Add to Cart
Fetuin, Fetal Bovine Serum is a liver-secreted 64 kDa plasma glycoprotein isolated from fetal bovine serum. It inhibits trypsin activity and promotes cellular attachment, growth, and differentiation. It is often an important supplement to many serum-free media. This acidic glycoprotein features three N-linked and three O-linked oligosaccharide chains, with terminal sugar residues rich in sialic acid (N-acetylneuraminic acid), contributing to its net negative charge.
  • Inhibits trypsin activity
  • Promotes cellular attachment
  • Promotes growth
  • Promotes differentiation
  • Important supplement to many serum-free media
  • Acidic glycoprotein with N-linked and O-linked oligosaccharide chains
  • Terminal sugar residues rich in sialic acid