Learn More
Medchemexpress LLC Fetuin, Fetal Bovine Serum | 9014-81-7 | 95% | 64 kDa | 500 MG
SDP
Catalog No. 5000422959
Encompass_Preferred
Fetuin, Fetal Bovine Serum is a liver-secreted 64 kDa plasma glycoprotein isolated from fetal bovine serum. It inhibits trypsin activity and promotes cellular attachment, growth, and differentiation. It is often an important supplement to many serum-free media.
  • Inhibits trypsin activity
  • Promotes cellular attachment, growth, and differentiation
  • Important supplement to many serum-free media
  • Acidic glycoprotein with three N-linked and three O-linked oligosaccharide chains
  • Terminal sugar residues are rich in sialic acid, contributing to its net negative charge
Catalog No. 50-004-22959 Supplier Medchemexpress LLC Supplier No. HYP2352500MG
Fetuin, Fetal Bovine Serum is a liver-secreted 64 kDa plasma glycoprotein isolated from fetal bovine serum. It inhibits trypsin activity and promotes cellular attachment, growth, and differentiation. It is often an important supplement to many serum-free media.
  • Inhibits trypsin activity
  • Promotes cellular attachment, growth, and differentiation
  • Important supplement to many serum-free media
  • Acidic glycoprotein with three N-linked and three O-linked oligosaccharide chains
  • Terminal sugar residues are rich in sialic acid, contributing to its net negative charge