Learn More
Selleck Chemical LLC Fetuin Fetal Bovine Serum P1243-25MG
SDP
Supplier:  Selleck Chemical LLC P124325MG
Encompass_Preferred
Fetuin Fetal Bovine Serum a prevalent protein found in fetal bovine plasma is the bovine equivalent of human 2HS glycoprotein ( 2HS) commonly used as a supplement in various serum-free media It inhibits trypsin activity and enhances cellular attachment growth and differentiation in culture systems
Catalog No. 50-000-39957