Learn More
QA Bio Inc Fetuin O glycan library
Catalog No. 502236574
Encompass
The fetuin O-linked glycan library in this product have been released from 50 µgs of a fetuin standard that is purified from fetal calf serum resulting in mixture of bi-, tri-, and tetra-antennary glycan standards with variable sialylation. A fetuin N-linked glycan library (CLIBN-FETUIN-01) and a fetuin glycoprotein (GCP-FET-05) are also available.
This item is not returnable. View return policy
Catalog No. 50-223-6574 Supplier QA Bio Inc Supplier No. CLIBOFETUIN01
The fetuin O-linked glycan library in this product have been released from 50 µgs of a fetuin standard that is purified from fetal calf serum resulting in mixture of bi-, tri-, and tetra-antennary glycan standards with variable sialylation. A fetuin N-linked glycan library (CLIBN-FETUIN-01) and a fetuin glycoprotein (GCP-FET-05) are also available.