Learn More
ACROBiosystems Recombinant Protein;Human Fetuin A / FETUA Protein;HEK293;1MG;FEA,AHSG,FETUA,Fetuin A,Fetuin-A,FetuinA,A2HS,AHS,FETUA,HSGA
Supplier:  ACROBiosystems FEAH52211MG
Encompass_Preferred
Recombinant Protein;Human Fetuin A / FETUA Protein;1MG;Human Fetuin A, His Tag (FEA-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Val 367 (Accession # NP_001613).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 38.2 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7917