Learn More
ACROBiosystems Recombinant Protein;Human Fetuin B / FETUA Protein;HEK293;100UG;FEB,FETUB,16G2,Gugu,Fetuin-B,Fetuin B,FetuinB
Supplier:  ACROBiosystems FEBH52H7100UG
Encompass_Preferred
Recombinant Protein;Human Fetuin B / FETUA Protein;100UG;Human Fetuin B, His Tag (FEB-H52H7) is expressed from human 293 cells (HEK293). It contains AA Cys 16 - Pro 382 (Accession # AAH69670 ).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 42.4 kDa. The protein migrates as 48-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8828