Learn More
Aviva Systems Biology Corp FEV Antibody - N-terminal region (ARP57889_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57889_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FEV. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human FEV. Alternate names: HSRNAFEV, PET-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-409