Learn More
Shockwatch Div Mri FeverScan Forehead Thermometer
SDP
Catalog No. NC2598434
Encompass_Preferred
FeverScan products utilize thermochromic liquid crystals for the measurement of clinical temperature trends in the health-care environment. FeverScan thermometers are composed of a series of discrete temperature events, each event shows a green color as the indicated temperature is attained. The color changes from tan to green to blue as the temperature increases. •94-105°F / 34 – 41°C•Easy to Read•Cost Effective•Watch for Hypo/Hyperthermia •Pre and Post-Op Surveillance •Hospital and Home Use •Disposable, Latex-Free •Core Adjusted Temperature•100 pieces / case
Catalog No. NC2598434 Supplier Shockwatch Div Mri Supplier No. P492
FeverScan products utilize thermochromic liquid crystals for the measurement of clinical temperature trends in the health-care environment. FeverScan thermometers are composed of a series of discrete temperature events, each event shows a green color as the indicated temperature is attained. The color changes from tan to green to blue as the temperature increases. •94-105°F / 34 – 41°C•Easy to Read•Cost Effective•Watch for Hypo/Hyperthermia •Pre and Post-Op Surveillance •Hospital and Home Use •Disposable, Latex-Free •Core Adjusted Temperature•100 pieces / case