Learn More
Shockwatch Div Mri FeverScan Forehead Thermometer
SDP
Catalog No. NC2598441
Encompass_Preferred
FeverScan products utilize thermochromic liquid crystals for the measurement of clinical temperature trends in the health-care environment. FeverScan thermometers are composed of a series of discrete temperature events, each event shows a green color as the indicated temperature is attained. The color changes from tan to green to blue as the temperature increases. •35-40°C•Easy to Read•Cost Effective•Watch for Hypo/Hyperthermia •Pre and Post-Op Surveillance •Hospital and Home Use •Disposable, Latex-Free •Core Adjusted Temperature•500 pieces / case (5 boxes of 100)
Catalog No. NC2598441 Supplier Shockwatch Div Mri Supplier No. P431
FeverScan products utilize thermochromic liquid crystals for the measurement of clinical temperature trends in the health-care environment. FeverScan thermometers are composed of a series of discrete temperature events, each event shows a green color as the indicated temperature is attained. The color changes from tan to green to blue as the temperature increases. •35-40°C•Easy to Read•Cost Effective•Watch for Hypo/Hyperthermia •Pre and Post-Op Surveillance •Hospital and Home Use •Disposable, Latex-Free •Core Adjusted Temperature•500 pieces / case (5 boxes of 100)