Learn More
Cayman Chemical FexapotIdetrIfluoroacetAT 10mg
SDP
Catalog No. 502942697
Encompass_Preferred
A peptide with anticancer activity; induces apoptosis in LNCaP prostate cancer cells at 2.5 mg/ml; reduces the numbers of prostate glandular epithelial cells without affecting other cell types and decreases prostate volume in rats when administered intraprostatically at 0.6 mg/animal per week
Catalog No. 50-294-2697 Supplier Cayman Chemical Supplier No. 4071610 MG
A peptide with anticancer activity; induces apoptosis in LNCaP prostate cancer cells at 2.5 mg/ml; reduces the numbers of prostate glandular epithelial cells without affecting other cell types and decreases prostate volume in rats when administered intraprostatically at 0.6 mg/animal per week