Learn More
Cayman Chemical Fexapotide (trifluoroacetate), Purity: ≥95%, CAS: 1609252-56-3, 50 mg
SDP
Catalog No. 502943717
Encompass_Preferred
A peptide with anticancer activity; induces apoptosis in LNCaP prostate cancer cells at 2.5 mg/ml; reduces the numbers of prostate glandular epithelial cells without affecting other cell types and decreases prostate volume in rats when administered intraprostatically at 0.6 mg/animal per week
Catalog No. 50-294-3717 Supplier Cayman Chemical Supplier No. 4071650 MG
A peptide with anticancer activity; induces apoptosis in LNCaP prostate cancer cells at 2.5 mg/ml; reduces the numbers of prostate glandular epithelial cells without affecting other cell types and decreases prostate volume in rats when administered intraprostatically at 0.6 mg/animal per week