Learn More
Medchemexpress LLC HY-10912 10mg Medchemexpress, Fexaramine CAS:574013-66-4 Purity:>98%
SDP
Catalog No. 502080765
Encompass_Preferred
Medchemexpress, HY-10912 10mg Fexaramine CAS:574013-66-4 Fexaramine is a potent and selective FXR agonist with an EC50 of 25 nM. Fexaramine has no activity against hRXRα, hPPARαγδ, mPXR, hPXR, hLXRα, hTRβ, hRARβ, mCAR, mERRγ, and hVDR receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0765 Supplier Medchemexpress LLC Supplier No. HY1091210MG
Medchemexpress, HY-10912 10mg Fexaramine CAS:574013-66-4 Fexaramine is a potent and selective FXR agonist with an EC50 of 25 nM. Fexaramine has no activity against hRXRα, hPPARαγδ, mPXR, hPXR, hLXRα, hTRβ, hRARβ, mCAR, mERRγ, and hVDR receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.