Learn More
Cayman Chemical Fexinidazole 10MG
SDP
Catalog No. 502965925
Encompass_Preferred
Fexinidazole is an antiparasitic agent; active against 15 strains of T. brucei, including T. b. rhodesiense, T. b. brucei, and T. b. gambiense (IC50s = 0.95-3.31 µM); inhibits the growth of L. donovani at the promastigote and axenic amastigote stages (EC50s = 5.6 and 2.8 µM, respectively) ; parasitic burden and increases survival in a mouse model of infection with benznidazole-susceptible, -partially resistant, or -resistant T. cruzi at doses ranging from 50-300 mg/kg per day
Catalog No. 50-296-5925 Supplier Cayman Chemical Supplier No. 1839310 MG
Fexinidazole is an antiparasitic agent; active against 15 strains of T. brucei, including T. b. rhodesiense, T. b. brucei, and T. b. gambiense (IC50s = 0.95-3.31 µM); inhibits the growth of L. donovani at the promastigote and axenic amastigote stages (EC50s = 5.6 and 2.8 µM, respectively) ; parasitic burden and increases survival in a mouse model of infection with benznidazole-susceptible, -partially resistant, or -resistant T. cruzi at doses ranging from 50-300 mg/kg per day