Learn More
Medchemexpress LLC HY-13801 5mg Medchemexpress, Fexinidazole CAS:59729-37-2 Purity:>98%
SDP
Catalog No. 502081515
Encompass_Preferred
Medchemexpress, HY-13801 5mg Fexinidazole CAS:59729-37-2 Fexinidazole (HOE 239) is an orally active, potent nitroimidazole antitrypanosomal drug. Fexinidazole shows trypanocidal activity against T. brucei subspecies and strains with IC50s of 0.7-3.3 μM (0.2-0.9 μg/ml). Fexinidazol has the potential for human sleeping sickness (HAT) caused by infection with T. brucei. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1515 Supplier Medchemexpress LLC Supplier No. HY138015MG
Medchemexpress, HY-13801 5mg Fexinidazole CAS:59729-37-2 Fexinidazole (HOE 239) is an orally active, potent nitroimidazole antitrypanosomal drug. Fexinidazole shows trypanocidal activity against T. brucei subspecies and strains with IC50s of 0.7-3.3 μM (0.2-0.9 μg/ml). Fexinidazol has the potential for human sleeping sickness (HAT) caused by infection with T. brucei. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.