Learn More
Medchemexpress LLC Fexofenadine | 83799-24-0 | >99.8% | 50 MG
SDP
Catalog No. 5000408144
Encompass_Preferred
Fexofenadine (MDL-16455) is an orally active and nonsedative H1 receptor antagonist. It is used in allergic rhinitis and chronic idiopathic urticarial research. Fexofenadine inhibits the expression of IL-6 protein in nasal fibroblasts in a dose-dependent manner and blocks phosphorylated p38 activation in histamine-induced nasal fibroblasts. In animal models, Fexofenadine hydrochloride has been shown to suppress both eosinophilia and systemic anaphylaxis in mice infected with *T. spiralis*.
  • Orally active and nonsedative H1 receptor antagonist.
  • Useful in allergic rhinitis and chronic idiopathic urticarial research.
  • Inhibits IL-6 protein expression in nasal fibroblasts.
  • Blocks phosphorylated p38 activation in histamine-induced nasal fibroblasts.
  • Suppresses eosinophilia and systemic anaphylaxis in animal models.
Catalog No. 50-004-08144 Supplier Medchemexpress LLC Supplier No. HYB080150MG
Fexofenadine (MDL-16455) is an orally active and nonsedative H1 receptor antagonist. It is used in allergic rhinitis and chronic idiopathic urticarial research. Fexofenadine inhibits the expression of IL-6 protein in nasal fibroblasts in a dose-dependent manner and blocks phosphorylated p38 activation in histamine-induced nasal fibroblasts. In animal models, Fexofenadine hydrochloride has been shown to suppress both eosinophilia and systemic anaphylaxis in mice infected with *T. spiralis*.
  • Orally active and nonsedative H1 receptor antagonist.
  • Useful in allergic rhinitis and chronic idiopathic urticarial research.
  • Inhibits IL-6 protein expression in nasal fibroblasts.
  • Blocks phosphorylated p38 activation in histamine-induced nasal fibroblasts.
  • Suppresses eosinophilia and systemic anaphylaxis in animal models.