Learn More
Medchemexpress LLC Fexofenadine-d6 | 548783-71-7 | 99.3% | 1 MG
SDP
Catalog No. 5000409188
Encompass_Preferred
Fexofenadine-d6 (MDL-16455-d6) is a deuterium-labeled form of Fexofenadine, which acts as an antihistamine pharmaceutical agent. This compound is intended for research use only. It can be utilized as a tracer and as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is used for quantitation during drug development and has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-09188 Supplier Medchemexpress LLC Supplier No. HYB0801S1MG
Fexofenadine-d6 (MDL-16455-d6) is a deuterium-labeled form of Fexofenadine, which acts as an antihistamine pharmaceutical agent. This compound is intended for research use only. It can be utilized as a tracer and as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is used for quantitation during drug development and has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs