Learn More
Apexbio Technology LLC Fexofenadine HCl 153439-40-8 50mg
SDP
Catalog No. 5000089107
Encompass_Preferred
Fexofenadine HCl (CAS 153439-40-8) is a non-sedating antihistamine that selectively antagonizes the histamine H1 receptor It inhibits H1 receptor-mediated signaling with an IC50 of 246 nM indicating potent receptor binding in vitro Fexofenadine is employed in biomedical research to investigate histaminergic pathways allergic responses and receptor pharmacology facilitating the study of H1 receptor antagonism in cellular and molecular models
Catalog No. 50-000-89107 Supplier Apexbio Technology LLC Supplier No. B157350
Fexofenadine HCl (CAS 153439-40-8) is a non-sedating antihistamine that selectively antagonizes the histamine H1 receptor It inhibits H1 receptor-mediated signaling with an IC50 of 246 nM indicating potent receptor binding in vitro Fexofenadine is employed in biomedical research to investigate histaminergic pathways allergic responses and receptor pharmacology facilitating the study of H1 receptor antagonism in cellular and molecular models