Learn More
Medchemexpress LLC Fexofenadine hydrochloride | 153439-40-8 | 99.9% | 5 MG
SDP
Catalog No. 5000408168
Encompass_Preferred
Fexofenadine hydrochloride is an orally active and nonsedative H1 receptor antagonist. It can be used in allergic rhinitis and chronic idiopathic urticarial research.
  • Orally active
  • Nonsedative H1 receptor antagonist
  • Can be used in allergic rhinitis research
  • Can be used in chronic idiopathic urticarial research
  • Inhibits the expression of IL-6 protein in nasal fibroblasts in a dose-dependent manner
  • Blocks phosphorylated p38 activation in histamine-induced nasal fibroblasts
  • Suppresses eosinophilia and systemic anaphylaxis in T. spiralis infected mice
Catalog No. 50-004-08168 Supplier Medchemexpress LLC Supplier No. HYB0801A5MG
Fexofenadine hydrochloride is an orally active and nonsedative H1 receptor antagonist. It can be used in allergic rhinitis and chronic idiopathic urticarial research.
  • Orally active
  • Nonsedative H1 receptor antagonist
  • Can be used in allergic rhinitis research
  • Can be used in chronic idiopathic urticarial research
  • Inhibits the expression of IL-6 protein in nasal fibroblasts in a dose-dependent manner
  • Blocks phosphorylated p38 activation in histamine-induced nasal fibroblasts
  • Suppresses eosinophilia and systemic anaphylaxis in T. spiralis infected mice