Learn More
Medchemexpress LLC Fexofenadine (hydrochloride) | 153439-40-8 | 99.9% | 50 MG
SDP
Catalog No. 5000409497
Encompass_Preferred
Fexofenadine hydrochloride is an orally active and nonsedative H1 receptor antagonist. It can be used in allergic rhinitis and chronic idiopathic urticarial research. In vitro studies show that Fexofenadine inhibits IL-6 protein expression and blocks phosphorylated p38 activation in histamine-induced nasal fibroblasts. In vivo, it suppresses eosinophilia and systemic anaphylaxis in T. spiralis-infected mice.
  • Orally active H1 receptor antagonist
  • Nonsedative properties
  • Inhibits IL-6 protein expression in nasal fibroblasts
  • Blocks phosphorylated p38 activation in histamine-induced nasal fibroblasts
  • Suppresses eosinophilia and systemic anaphylaxis in relevant animal models
Catalog No. 50-004-09497 Supplier Medchemexpress LLC Supplier No. HYB0801A50MG
Fexofenadine hydrochloride is an orally active and nonsedative H1 receptor antagonist. It can be used in allergic rhinitis and chronic idiopathic urticarial research. In vitro studies show that Fexofenadine inhibits IL-6 protein expression and blocks phosphorylated p38 activation in histamine-induced nasal fibroblasts. In vivo, it suppresses eosinophilia and systemic anaphylaxis in T. spiralis-infected mice.
  • Orally active H1 receptor antagonist
  • Nonsedative properties
  • Inhibits IL-6 protein expression in nasal fibroblasts
  • Blocks phosphorylated p38 activation in histamine-induced nasal fibroblasts
  • Suppresses eosinophilia and systemic anaphylaxis in relevant animal models