Learn More
Abcam Fe x ofenadine hydrochloride, H1 receptor antagonist, 50MG
Catalog No. 502707292
Encompass_Preferred
MW 538.13 g/mol, Purity >99%. Selective H₁ receptor antagonist (Ki = 10 nM). Does not readily cross the blood-brain barrier. Orally active antiallergenic.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7292 Supplier Abcam Supplier No. AB12076550MG
MW 538.13 g/mol, Purity >99%. Selective H₁ receptor antagonist (Ki = 10 nM). Does not readily cross the blood-brain barrier. Orally active antiallergenic.
The product is subject to the following: Abcam Restricted Use Statement