Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fexofenadine Related Compound C, MilliporeSigma™ Supelco™

Catalog No. 111002279 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2279 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2279 Supplier MilliporeSigma Supelco Supplier No. 127044625MG
Add to Cart
Add to Cart
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 185066-37-9 (free base)
Purity Grade Notes These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind. ​
Physical Form Neat
Linear Formula C31H39 NO2 · HCl
Synonym 4-[4-(Hydroxydiphenylmethyl)piperidinyl]-1-(p-isopropylphenyl)butane-1-ol hydrochloride
Type USP Reference Standard
Quantity 25 mg
Formula Weight 494.11
Grade Pharmaceutical Primary Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.