Learn More
Cayman Chemical Fexofenadine (hydrochloride), Purity: ≥98%, CAS: 153439-40-8, 100 mg
SDP
Catalog No. 502965007
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 10 nM); reverses acetyl-β-methylcholine-induced contraction of isolated rat tracheal strips; inhibits expression of IL-8 in TNF-α-stimulated HCT116 and COLO 205 cells; reduces severity of colitis and phospho-IκB kinase activation in a mouse model of DSS-induced colitis
Catalog No. 50-296-5007 Supplier Cayman Chemical Supplier No. 18191100 MG
A histamine H1 receptor antagonist (Ki = 10 nM); reverses acetyl-β-methylcholine-induced contraction of isolated rat tracheal strips; inhibits expression of IL-8 in TNF-α-stimulated HCT116 and COLO 205 cells; reduces severity of colitis and phospho-IκB kinase activation in a mouse model of DSS-induced colitis