Learn More
Cayman Chemical FexofenadInhydrochlorIde 10mg
SDP
Catalog No. 502966548
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 10 nM); reverses acetyl-β-methylcholine-induced contraction of isolated rat tracheal strips; inhibits expression of IL-8 in TNF-α-stimulated HCT116 and COLO 205 cells; reduces severity of colitis and phospho-IκB kinase activation in a mouse model of DSS-induced colitis
Catalog No. 50-296-6548 Supplier Cayman Chemical Supplier No. 1819110 MG
A histamine H1 receptor antagonist (Ki = 10 nM); reverses acetyl-β-methylcholine-induced contraction of isolated rat tracheal strips; inhibits expression of IL-8 in TNF-α-stimulated HCT116 and COLO 205 cells; reduces severity of colitis and phospho-IκB kinase activation in a mouse model of DSS-induced colitis