Learn More
Medchemexpress LLC Fezagepras (sodium) | 1254472-97-3 | 99.8% | 228.26 g/mol | C13H17NaO2 | 1 ML
SDP
Catalog No. 5000211295
Encompass_Preferred
Fezagepras sodium (setogepram sodium) is a research compound used in studies of GPR40/GPR84-related biology with reported anti-fibrotic and anti-inflammatory effects. It is supplied as solids and as a 10 mM solution in DMSO (1 mL), and is accompanied by manufacturer COA and SDS documents. Recommended storage is sealed and dry; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • High purity (≈99.8%) for research and analytical use.
  • Available as ready-to-use 10 mM solution in DMSO and as solid forms.
  • COA and SDS documentation support traceability and safe handling.
  • Storage recommendations help preserve compound stability.
  • Applicable for in vitro assays targeting GPR40/GPR84 and fibrosis models.
Catalog No. 50-002-11295 Supplier Medchemexpress LLC Supplier No. HY10077510MM 1ML
Fezagepras sodium (setogepram sodium) is a research compound used in studies of GPR40/GPR84-related biology with reported anti-fibrotic and anti-inflammatory effects. It is supplied as solids and as a 10 mM solution in DMSO (1 mL), and is accompanied by manufacturer COA and SDS documents. Recommended storage is sealed and dry; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • High purity (≈99.8%) for research and analytical use.
  • Available as ready-to-use 10 mM solution in DMSO and as solid forms.
  • COA and SDS documentation support traceability and safe handling.
  • Storage recommendations help preserve compound stability.
  • Applicable for in vitro assays targeting GPR40/GPR84 and fibrosis models.