Learn More
TARGETMOL CHEMICALS INC FEZAGEPRAS SODIUM 5MG
SDP
Catalog No. 502581461
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fezagepras sodium (Setogepram sodium salt) is an orally active GPR40 agonist and is an antagonist or inverse agonist for GPR84 with anti-fibrotic anti-inflammatory and anti-proliferative actions. purity: 99%
Catalog No. 50-258-1461 Supplier TARGETMOL CHEMICALS INC Supplier No. T123755MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fezagepras sodium (Setogepram sodium salt) is an orally active GPR40 agonist and is an antagonist or inverse agonist for GPR84 with anti-fibrotic anti-inflammatory and anti-proliferative actions. purity: 99%