Learn More
Medchemexpress LLC HY-19632 5mg Medchemexpress, Fezolinetant CAS:1629229-37-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY196325MG
Encompass_Preferred
Medchemexpress, HY-19632 5mg Fezolinetant CAS:1629229-37-3 Fezolinetant is an antagonist of the neurokinin 3 receptor (NK3R), used for the treatment of menopausal hot flushes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2156