Learn More
Cayman Chemical Fezolinetant, Purity: ≥98%, CAS: 1629229-37-3, 10 mg
SDP
Catalog No. 502918907
Encompass_Preferred
An NK3 receptor antagonist (IC50 = 20 nM); decreases plasma levels of LH in castrated cynomolgus monkeys at 5 mg/kg; prevents increases in body weight and food intake but does not affect uterine weight or plasma levels of estradiol, FSH, progesterone, or testosterone in ovariectomized rats at 10 mg/kg; inhibits increases in skin temperature but not core temperature, indicating regulation of hot flash-like symptoms, in ovariectomized rats at 3 or 10 mg/kg
Catalog No. 50-291-8907 Supplier Cayman Chemical Supplier No. 3952310 MG
An NK3 receptor antagonist (IC50 = 20 nM); decreases plasma levels of LH in castrated cynomolgus monkeys at 5 mg/kg; prevents increases in body weight and food intake but does not affect uterine weight or plasma levels of estradiol, FSH, progesterone, or testosterone in ovariectomized rats at 10 mg/kg; inhibits increases in skin temperature but not core temperature, indicating regulation of hot flash-like symptoms, in ovariectomized rats at 3 or 10 mg/kg