Learn More
Imtakt PRESTO FF-C18, 150 x 3 mm
Catalog No. NC1669207
Encompass
This is the world’s first 2µ non-porous column. This extremely efficient material works very well for separation of large molecules like polymers, surfactants, protein and even DNA. It has a wide effective analysis range from 5kDa to 50 MDa. Since it does not have any pores,there is essentially no band broadening due to mass transfer effect and no size limit for the compounds that it can retain. We believe so strongly in the value of non-porous columns that we intend to develop an entire line of non-porous columns in the future.
Catalog No. NC1669207 Supplier Imtakt Supplier No. FF035
This is the world’s first 2µ non-porous column. This extremely efficient material works very well for separation of large molecules like polymers, surfactants, protein and even DNA. It has a wide effective analysis range from 5kDa to 50 MDa. Since it does not have any pores,there is essentially no band broadening due to mass transfer effect and no size limit for the compounds that it can retain. We believe so strongly in the value of non-porous columns that we intend to develop an entire line of non-porous columns in the future.