Learn More
Biolog Inc FF MICRO PLATE 10 PLATES/PK
SDP
Catalog No. NC9007685
Encompass
FF MicroPlate, Rapid Identification and Characterization Product, Application: Designed for Filamentous Fungi And Yeast
Catalog No. NC9007685 Supplier Biolog Inc Supplier No. 1006
FF MicroPlate, Rapid Identification and Characterization Product, Application: Designed for Filamentous Fungi And Yeast