Learn More
FFA Solution, Toluene Free, Reagents
Specifications
Specifications
| Chemical Name or Material | FFA Solution, Toluene Free |
| Color | Colorless to Pink |
| CAS | 67-63-0, 7732-18-5 |
| Boiling Point | 83°C |
| Synonym | Free Fatty Acid Isopropanol |
| Grade | Reagent |
| Packaging | Steel Pail |
| Specific Gravity | 0.781 |
| Quantity | 20 L |
| Sterility | Non-sterile |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.