Learn More
Biomatik Corporation Ffar1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11310
Encompass_Preferred
Ffar1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to FFAR1. The Ffar1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Free Fatty Acid Receptor 1 Protein (143-178Aa). Applications: ELISA, IHC, IF. Target Synonyms: FFAR1. Accession Number: O14842. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5367