Learn More
Aviva Systems Biology Corp FFAR2 Antibody - C-terminal region (ARP65165_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65165_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FFAR2. It was validated on Western Blot. . Alternate names: FFA2R, GPR43ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-288-445