Learn More
Eurofins Discoverx Ready-to-Assay FFA3/GPR41 Free Fatty Acid Receptor Frozen Cells, 2 Vials of Cells. (HTS135RTA)
Catalog No. 502005953
Encompass
The Ready-to-Assay FFA3/GPR41 Free Fatty Acid Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. free fatty acids) based activation of FFAR3 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses FFAR3 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of FFAR3, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5953 Supplier Eurofins Discoverx Supplier No. HTS135RTA
The Ready-to-Assay FFA3/GPR41 Free Fatty Acid Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. free fatty acids) based activation of FFAR3 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses FFAR3 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of FFAR3, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.