Learn More
Eurofins Discoverx ChemiScreen FFAR3/GPR41 Free Fatty Acid Receptor Stable Cell Line, 2 vials. (HTS135C)
Catalog No. 502005952
Encompass
The ChemiScreen FFAR3/GPR41 Free Fatty Acid Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of FFAR3 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses FFAR3 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of FFAR3, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5952 Supplier Eurofins Discoverx Supplier No. HTS135C
Add to Cart
Add to Cart
Request a Quote
The ChemiScreen FFAR3/GPR41 Free Fatty Acid Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of FFAR3 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses FFAR3 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of FFAR3, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.