Learn More
Cayman Chemical FFAR4 (GPR120) (Internal) Polyclonal Antibody, 1 ea
SDP
Catalog No. 502971359
Encompass_Preferred
Immunogen: Synthetic peptide from the internal region of human FFAR4 (GPR120) • Host: Rabbit • Species Reactivity: (+) Human • Application(s): ELISA and WB
Catalog No. 50-297-1359 Supplier Cayman Chemical Supplier No. 149041 EA
Immunogen: Synthetic peptide from the internal region of human FFAR4 (GPR120) • Host: Rabbit • Species Reactivity: (+) Human • Application(s): ELISA and WB