Learn More
Bachem H-Phe-Phe-Phe-OH
SDP
Supplier:  Bachem 4011114.1000
Encompass_Preferred
H-Phe-Phe-Phe-OH, 2578-81-6, C27H29N3O4, Mr 459.55, 1g. FFF is a bitter-tasting peptide due to its hydrophobicity. As diphenylalanine, triphenylalanine forms ordered nanostructures.
Catalog No. 50-194-7518