Learn More
Medchemexpress LLC FFN 102 mesylate | 1883548-92-2 | 99.0% | 335.76 | 50 MG
SDP
Catalog No. 5000344109
Encompass_Preferred
FFN 102 mesylate is a synthetic biogenic neurotransmitter analogue, characterized by its pH-dependent fluorescence and electrical activity. It is intended for research use only. For proper storage, it should be kept at -20°C in sealed storage, away from moisture. When dissolved, stock solutions in DMSO or water have specific storage conditions: -80°C for 6 months or -20°C for 1 month (sealed, away from moisture).
  • Requires ultrasonic treatment for dissolution in DMSO and water.
  • Hygroscopic nature of DMSO impacts product solubility.
  • Dilute aqueous stock solutions to working solutions, then filter and sterilize with a 0.22 μm filter before use.
Catalog No. 50-003-44109 Supplier Medchemexpress LLC Supplier No. HY11033710MG
FFN 102 mesylate is a synthetic biogenic neurotransmitter analogue, characterized by its pH-dependent fluorescence and electrical activity. It is intended for research use only. For proper storage, it should be kept at -20°C in sealed storage, away from moisture. When dissolved, stock solutions in DMSO or water have specific storage conditions: -80°C for 6 months or -20°C for 1 month (sealed, away from moisture).
  • Requires ultrasonic treatment for dissolution in DMSO and water.
  • Hygroscopic nature of DMSO impacts product solubility.
  • Dilute aqueous stock solutions to working solutions, then filter and sterilize with a 0.22 μm filter before use.