Learn More
Medchemexpress LLC FFN 206 dihydrochloride | 1883548-88-6 | 99.5% | 291.17 | 25 MG
SDP
Catalog No. 5000345905
Encompass_Preferred
FFN 206 dihydrochloride is a fluorescent probe utilized as an excellent Vesicular Monoamine Transporter 2 (VMAT2) substrate with an apparent Km of 1.16 μM. It can detect VMAT2 activity in intact cells using fluorescence microscopy, localizing to VMAT2-expressing acidic compartments without apparent labeling of other organelles. This probe is for research use only and not sold to patients.
  • Detects VMAT2 activity in intact cells
  • Subcellular localization to VMAT2-expressing acidic compartments
Catalog No. 50-003-45905 Supplier Medchemexpress LLC Supplier No. 25MGHY110334
FFN 206 dihydrochloride is a fluorescent probe utilized as an excellent Vesicular Monoamine Transporter 2 (VMAT2) substrate with an apparent Km of 1.16 μM. It can detect VMAT2 activity in intact cells using fluorescence microscopy, localizing to VMAT2-expressing acidic compartments without apparent labeling of other organelles. This probe is for research use only and not sold to patients.
  • Detects VMAT2 activity in intact cells
  • Subcellular localization to VMAT2-expressing acidic compartments